Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals
Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
1 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 217120025
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
Add to cart
Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals
Catalog number217120025
Price (USD)/ Each
102.65
Online exclusive
114.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS144-55-8
IUPAC Namesodium hydrogen carbonate
Molecular FormulaCHNaO3
InChI KeyUIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES[Na+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.015 %
Chloride (Cl)=<100 ppm
Ammonium (NH4)=<5 ppm
Total sulfur (S)=<10 ppm
Copper (Cu)=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only