Thermo Scientific Chemicals

Toluene-d8, for NMR, 100.0 atom % D, packaged in 0.50 ml ampoules, Thermo Scientific Chemicals

Catalog number: 217440050
5 mL, Ampule, Each
Thermo Scientific Chemicals

Toluene-d8, for NMR, 100.0 atom % D, packaged in 0.50 ml ampoules, Thermo Scientific Chemicals

Catalog number: 217440050
5 mL, Ampule, Each
Quantity
Packaging
Catalog number: 217440050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2037-26-5
IUPAC Name
1-(²H₃)methyl(²H₅)benzene
Molecular Formula
C7H8
InChI Key
YXFVVABEGXRONW-JGUCLWPXSA-N
SMILES
[2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])[2H]
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
D-Enrichment
>=99.95 %
Water
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text