Aluminum chloride, 98.5%, extra pure, anhydrous, powder
Aluminum chloride, 98.5%, extra pure, anhydrous, powder
Aluminum chloride, 98.5%, extra pure, anhydrous, powder
Aluminum chloride, 98.5%, extra pure, anhydrous, powder
Aluminum chloride, 98.5%, extra pure, anhydrous, powder
Thermo Scientific Chemicals

Aluminum chloride, 98.5%, extra pure, anhydrous, powder

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 217461000
Price (USD)
51.65
Online Exclusive
57.50
Save 5.85 (10%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.65
Online Exclusive
57.50
Save 5.85 (10%)
Each
Add to cart
Chemical Identifiers
CAS7446-70-0
IUPAC Namealuminium(3+) trichloride
Molecular FormulaAlCl3
InChI KeyVSCWAEJMTAWNJL-UHFFFAOYSA-K
SMILES[Al+3].[Cl-].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.2 % (in water)
Iron (Fe)=<100 ppm
Nickel (Ni)=<20 ppm
Appearance (Color)White to yellow to grey
Titration Argentometric>=98.0 %
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Lewis acid
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link