Thermo Scientific Chemicals

Aluminum chloride hexahydrate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 217470100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Aluminum chloride hexahydrate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 217470100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 217470100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7784-13-6
IUPAC Name
aluminium(3+) hexahydrate trichloride
Molecular Formula
AlCl3H12O6
InChI Key
JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
Appearance (Color)
White to yellow
Appearance (Form)
Adhering powder or crystals
Titration Argentometric
>=98.5 %
Water
42 to 48 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text