Barium carbonate, 99+%, for analysis, Thermo Scientific Chemicals
Barium carbonate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium carbonate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number 217490025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS513-77-9
IUPAC Namebarium(2+) carbonate
Molecular FormulaCBaO3
InChI KeyAYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES[Ba++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration Complexometric>=99.0 %
Heavy metals (as Pb)=<10 ppm
Insoluble matter=<0.015 % (in dilute HCl)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only