Thermo Scientific Chemicals

Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals

Catalog number: 217500100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Bis(triphenylphosphine)nickel(II)chloride, 98%, Thermo Scientific Chemicals

Catalog number: 217500100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 217500100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14264-16-5
IUPAC Name
dichloronickelbis(ylium); bis(triphenylphosphane)
Molecular Formula
C36H30Cl2NiP2
InChI Key
VHQPRJDILRYZBG-UHFFFAOYSA-L
SMILES
Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Dark green to dark grey
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Elemental Analysis
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text