Chemical Identifiers
CAS14264-16-5
IUPAC Namedichloronickelbis(ylium); bis(triphenylphosphane)
Molecular FormulaC36H30Cl2NiP2
InChI KeyVHQPRJDILRYZBG-UHFFFAOYSA-L
SMILESCl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more