Thermo Scientific Chemicals

Lead(IV) oxide, 97+%, extra pure, Thermo Scientific Chemicals

Catalog number: 217535000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Lead(IV) oxide, 97+%, extra pure, Thermo Scientific Chemicals

Catalog number: 217535000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 217535000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1309-60-0
IUPAC Name
plumbanedione
Molecular Formula
O2Pb
InChI Key
YADSGOSSYOOKMP-UHFFFAOYSA-N
SMILES
O=[Pb]=O
Appearance (Color)
Brown to black
Titration with Na2S2O3
>=97.0 %
Sulfate (SO4)
=<0.005 %
Iron (Fe)
=<0.02 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text