(S)-(+)-1,3-Butanediol, 98%, Thermo Scientific Chemicals
(S)-(+)-1,3-Butanediol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(+)-1,3-Butanediol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 217970050
Price (USD)/ Each
342.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
342.00
Add to cart
(S)-(+)-1,3-Butanediol, 98%, Thermo Scientific Chemicals
Catalog number217970050
Price (USD)/ Each
342.00
-
Add to cart
Chemical Identifiers
CAS24621-61-2
IUPAC Namebutane-1,3-diol
Molecular FormulaC4H10O2
InChI KeyPUPZLCDOIYMWBV-UHFFFAOYNA-N
SMILESCC(O)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4385 to 1.4405 (20°C, 589 nm)
Appearance (Form)Viscous liquid
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only