Thermo Scientific Chemicals

L(+)-α-Phenylglycine, 99%

Catalog number: 217990250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

L(+)-α-Phenylglycine, 99%

Catalog number: 217990250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 217990250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2935-35-5
IUPAC Name
2-amino-2-phenylacetic acid
Molecular Formula
C8H9NO2
InChI Key
ZGUNAGUHMKGQNY-UHFFFAOYNA-N
SMILES
NC(C(O)=O)C1=CC=CC=C1
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Titration with HClO4
>=98.5 %
Specific optical rotation
+154° to +160° (20°C, 589 nm) (c=1, 1 M HCl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text