Thermo Scientific Chemicals

7-Amino-4-methylcoumarin, 98%

Catalog number: 218060010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

7-Amino-4-methylcoumarin, 98%

Catalog number: 218060010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 218060010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
26093-31-2
IUPAC Name
7-amino-4-methyl-2H-chromen-2-one
Molecular Formula
C10H9NO2
InChI Key
GLNDAGDHSLMOKX-UHFFFAOYSA-N
SMILES
CC1=CC(=O)OC2=CC(N)=CC=C12
HPLC
>=97.5 %
Appearance (Color)
Yellow to beige to brown
Infrared spectrum
Conforms
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text