Thermo Scientific Chemicals

Tetraheptylammonium bromide, 99%, Thermo Scientific Chemicals

Catalog number: 218161000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetraheptylammonium bromide, 99%, Thermo Scientific Chemicals

Catalog number: 218161000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 218161000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4368-51-8
IUPAC Name
tetraheptylazanium bromide
Molecular Formula
C28H60BrN
InChI Key
YQIVQBMEBZGFBY-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC
Titration Argentometric
>=98.5 %
Appearance (Color)
White
Infrared spectrum
Conforms
Melting point
89°C to 91°C
Appearance (Form)
Flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text