Thermo Scientific Chemicals

Trimethyl orthoacetate, 98+%, Thermo Scientific Chemicals

Catalog number: 218185000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Trimethyl orthoacetate, 98+%, Thermo Scientific Chemicals

Catalog number: 218185000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 218185000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1445-45-0
IUPAC Name
1,1,1-trimethoxyethane
Molecular Formula
C5H12O3
InChI Key
HDPNBNXLBDFELL-UHFFFAOYSA-N
SMILES
COC(C)(OC)OC
Infrared spectrum
Conforms
GC
>=98.0 %
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Refractive index
1.3870 to 1.3900 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text