Thermo Scientific Chemicals

Phenyl chloroformate, 99%, Thermo Scientific Chemicals

Catalog number: 218212500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Phenyl chloroformate, 99%, Thermo Scientific Chemicals

Catalog number: 218212500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 218212500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1885-14-9
IUPAC Name
phenyl carbonochloridate
Molecular Formula
C7H5ClO2
InChI Key
AHWALFGBDFAJAI-UHFFFAOYSA-N
SMILES
ClC(=O)OC1=CC=CC=C1
Infrared spectrum
Conforms
Refractive index
1.5100 to 1.5120 (20°C, 589 nm)
Appearance (Form)
Clear liquid
Color scale
=<50 APHA
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text