Thermo Scientific Chemicals

Calcium fluoride, 99%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 218255000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium fluoride, 99%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 218255000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 218255000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-75-5
IUPAC Name
calcium difluoride
Molecular Formula
CaF2
InChI Key
WUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES
[F-].[F-].[Ca++]
Assay
>=98.5 % (CaF2)
Appearance (Color)
White to off-white
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text