Lithium fluoride, 97%, for analysis, Thermo Scientific Chemicals
Lithium fluoride, 97%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium fluoride, 97%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 218271000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7789-24-4
IUPAC Namelithium(1+) fluoride
Molecular FormulaFLi
InChI KeyPQXKHYXIUOZZFA-UHFFFAOYSA-M
SMILES[Li+].[F-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Loss on drying0.4 % (200°C) (typical)
Sulfate (SO4)=<1 %
Carbonate (CO3)=<1 %
Potassium (K)=<0.07 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only