Thermo Scientific Chemicals

Lithium fluoride, 97%, for analysis, Thermo Scientific Chemicals

Catalog number: 218275000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Lithium fluoride, 97%, for analysis, Thermo Scientific Chemicals

Catalog number: 218275000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 218275000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-24-4
IUPAC Name
lithium(1+) fluoride
Molecular Formula
FLi
InChI Key
PQXKHYXIUOZZFA-UHFFFAOYSA-M
SMILES
[Li+].[F-]
Assay
>=96.0 %
Chloride (Cl)
=<0.07 %
Phosphate (PO4)
=<0.04 %
Iron (Fe)
=<0.03 %
Sodium (Na)
=<0.4 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text