Lithium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
Lithium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 218320010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10377-48-7
IUPAC Namedilithium(1+) sulfate
Molecular FormulaLi2O4S
InChI KeyINHCSSUBVCNVSK-UHFFFAOYSA-L
SMILES[Li+].[Li+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
Appearance (Form)Powder
Insoluble matter=<0.01 %
Appearance (Color)White
Titration after Ion exchange>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only