Thermo Scientific Chemicals

o-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 218480250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

o-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 218480250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 218480250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
615-28-1
IUPAC Name
dihydrogen benzene-1,2-diamine dichloride
Molecular Formula
C6H10Cl2N2
InChI Key
RIIWUGSYXOBDMC-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NC1=CC=CC=C1N
Appearance (Form)
Crystalline powder
Appearance (Color)
Light pink to purple
Titration Argentometric
>=99.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text