o-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals
o-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Packaging:
Glass bottle
Catalog number 218481000
Price (USD)/ Each
434.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
434.00
Add to cart
o-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals
Catalog number218481000
Price (USD)/ Each
434.00
-
Add to cart
Chemical Identifiers
IUPAC Namedihydrogen benzene-1,2-diamine dichloride
Molecular FormulaC6H10Cl2N2
InChI KeyRIIWUGSYXOBDMC-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NC1=CC=CC=C1N
Molecular Weight (g/mol)181.06
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Light pink to purple
Titration Argentometric>=99.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only