Cyclohexyl iodide, 98%, stabilized, Thermo Scientific Chemicals
Cyclohexyl iodide, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexyl iodide, 98%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 218490250
Price (USD)/ Each
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS626-62-0
IUPAC Nameiodocyclohexane
Molecular FormulaC6H11I
InChI KeyFUCOMWZKWIEKRK-UHFFFAOYSA-N
SMILESIC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5450 to 1.5490 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to yellow to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only