1-Iodohexane, 98%, stabilized, Thermo Scientific Chemicals
1-Iodohexane, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Iodohexane, 98%, stabilized, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number 218521000
Price (USD)/ Each
61.65
Online exclusive
68.40 
Save 6.75 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
68.40 
Save 6.75 (10%)
Add to cart
1-Iodohexane, 98%, stabilized, Thermo Scientific Chemicals
Catalog number218521000
Price (USD)/ Each
61.65
Online exclusive
68.40 
Save 6.75 (10%)
-
Add to cart
Chemical Identifiers
CAS638-45-9
IUPAC Name1-iodohexane
Molecular FormulaC6H13I
InChI KeyANOOTOPTCJRUPK-UHFFFAOYSA-N
SMILESCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4900 to 1.4940 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
StabilizerCopper
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only