SPDP (succinimidyl 3-(2-pyridyldithio)propionate)
SPDP (succinimidyl 3-(2-pyridyldithio)propionate)
Thermo Scientific™

SPDP (succinimidyl 3-(2-pyridyldithio)propionate)

Thermo Scientific Pierce SPDP is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavableRead more
Have Questions?
Catalog NumberQuantity
2185750 mg
Catalog number 21857
Price (USD)
221.00
Each
-
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (USD)
221.00
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce SPDP is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls.

Features of SPDP:

Reactive groups: NHS ester and pyridyldithiol
Reactive towards: amino and sulfhydryl groups
• Releases a detectable by-product when reacted with a free sulfhydryl group; by measuring the release of pyridine-2-thione at 343 nm, the reaction can be easily followed
• Disulfide bond in the spacer arm is readily cleaved by 10-50 mM DTT or TCEP at pH 8.5
• Spacer arm is also easily cleaved using reducing SDS-PAGE sample loading buffer
• Cleavable crosslinker allows separation of crosslinked products
• Water-insoluble (dissolve first in DMF or DMSO)
• SPDP crosslinker is membrane permeable, so crosslinking can be done inside the cells
• Compare to other varieties of SPDP-type reagents, including pegylated forms

Applications:
• Reversible crosslinking of proteins
• Adding sulfhydryl groups to proteins by attachment to lysine residues
• Protein crosslinking to create antibody-enzyme and other bioconjugates via a two-step reaction
• Attaching haptens to carrier proteins for antibody production
• Preparing immunotoxin conjugates

Specifications:
We manufacture SPDP to the highest specification to produce the most specific bioconjugates, ensure the integrity of your data and to provide you with the highest degree of consistency. Each lot of SPDP is tested to ensure it is >95% pure by quantitative NMR (the highest standard for crosslinker purity).

We offer three analogs of SPDP: the standard version (SPDP), a derivative with a longer spacer arm (LC-SPDP), and a sulfonated water-soluble variety (Sulfo-LC-SPDP). SPDP (N-Succinimidyl 3-(2-pyridyldithio)-propionate) and LC-SPDP (Succinimidyl 6-(3-[2-pyridyldithio]-propionamido)hexanoate) are heterobifunctional, thiol-cleavable and membrane permeable crosslinkers. They each contain an amine-reactive N-hydroxysuccinimide (NHS) ester that will react with lysine residues to form a stable amide bond. The other end of the spacer arm is terminated in the pyridyl disulfide group that will react with sulfhydryls to form a reversible disulfide bond.

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
Pierce™ Premium Grade SPDP
LC-SPDP (succinimidyl 6-(3(2-pyridyldithio)propionamido)hexanoate)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityYes
DescriptionSPDP
FormPowder
Labeling MethodChemical Labeling
Molecular Weight (g/mol)312.36
PEGylatedNo
Product LinePierce™
Quantity50 mg
Reactive MoietyNHS Ester, Pyridyldisulfide
Shipping ConditionAmbient
SolubilityDMF, DMSO
Spacer Arm Length6.8 Å
Water SolubleNo
Chemical ReactivityAmine-Sulfhydryl
CleavableBy Thiols
Crosslinker TypeHeterobifunctional
FormatStandard, Premium-grade
Product TypeCrosslinker
SpacerShort (<10 Å)
Unit SizeEach
Contents & Storage
Upon receipt store at -20°C protected from moisture.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can you provide the shelf-life for SPDP (succinimidyl 3-(2-pyridyldithio)propionate)?

SPDP (succinimidyl 3-(2-pyridyldithio)propionate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection