Chemical Identifiers
CAS458-37-7
IUPAC Name(1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one
Molecular FormulaC21H20O6
InChI KeyZIUSSTSXXLLKKK-KOBPDPAPSA-N
SMILESCOC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
View more