Thermo Scientific Chemicals

Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 98+%

Catalog number: 218582500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 98+%

Catalog number: 218582500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 218582500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
458-37-7
IUPAC Name
(1E,4Z,6E)-5-hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,4,6-trien-3-one
Molecular Formula
C21H20O6
InChI Key
ZIUSSTSXXLLKKK-KOBPDPAPSA-N
SMILES
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
Appearance (Color)
Orange
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
UV spectrophotometry
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text