Bacto™ Malt Extract
Bacto™ Malt Extract
Gibco™

Bacto™ Malt Extract

Bacto Malt Extract is the water-soluble portion of malted barley. Due to the extraction process, this animal origin-free supplement providesRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
218630500 g
21861010 kg
Catalog number 218630
Price (USD)/ Each
53.75
Add to cart
Quantity:
500 g
Price (USD)/ Each
53.75
Add to cart
Bacto Malt Extract is the water-soluble portion of malted barley. Due to the extraction process, this animal origin-free supplement provides a very high carbohydrate content to any medium. Bacto Malt Extract has been used successfully in microbial fermentation applications.

Specifications
Certifications/ComplianceLUO
FormPowder
Product TypeMalt Extract
Shelf Life1460 Days
FormatBottle
For Use With (Application)Microbial fermentation applications
Product LineBacto™
Quantity500 g
SourceAnimal origin-free, malted barley
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.

Share catalog number, name or link