Sodium tripolyphosphate, 85%, pure, Tech.
Sodium tripolyphosphate, 85%, pure, Tech.
Sodium tripolyphosphate, 85%, pure, Tech.
Sodium tripolyphosphate, 85%, pure, Tech.
Thermo Scientific Chemicals

Sodium tripolyphosphate, 85%, pure, Tech.

CAS: 7758-29-4 | Na5O10P3 | 367.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 218670025
Price (EUR)
232,05
Online Exclusive
273,00
Save 40,95 (15%)
Each
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
232,05
Online Exclusive
273,00
Save 40,95 (15%)
Each
Add to cart
Chemical Identifiers
CAS7758-29-4
IUPAC Namepentasodium bis(phosphonatooxy)phosphinate
Molecular FormulaNa5O10P3
InChI KeyHWGNBUXHKFFFIH-UHFFFAOYSA-I
SMILES[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Titration with HClO4>=80 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0467034Certificate of AnalysisNov 07, 2024218670025, 218675000
A0450777Certificate of AnalysisMay 02, 2023218670025, 218675000
A0392666Certificate of AnalysisDec 15, 2022218670025, 218675000
A0407019Certificate of AnalysisDec 13, 2022218670025, 218675000
A0438352Certificate of AnalysisJan 24, 2022218670025, 218675000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection