Thermo Scientific Chemicals

Cerium(III) nitrate hexahydrate, 99.5%, Thermo Scientific Chemicals

Catalog number: 218695000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Cerium(III) nitrate hexahydrate, 99.5%, Thermo Scientific Chemicals

Catalog number: 218695000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 218695000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-41-4
IUPAC Name
cerium(3+) hexahydrate trinitrate
Molecular Formula
CeH12N3O15
InChI Key
QQZMWMKOWKGPQY-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Assay
>=99.4 % (CeO2/TREO basis)
Calcium oxide (CaO)
=<50 ppm
Appearance (Form)
Powder or crystals and/or chunks
Appearance (Color)
Colorless to white
Iron(III)oxide (Fe2O3)
=<35 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text