Thermo Scientific Chemicals

Cesium sulfate, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 218701000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Cesium sulfate, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 218701000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 218701000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-54-9
IUPAC Name
dicaesium(1+) sulfate
Molecular Formula
Cs2O4S
InChI Key
FLJPGEWQYJVDPF-UHFFFAOYSA-L
SMILES
[Cs+].[Cs+].[O-]S([O-])(=O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Assay
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text