Lanthanum(III) nitrate hexahydrate, 98+%, Thermo Scientific Chemicals
Lanthanum(III) nitrate hexahydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum(III) nitrate hexahydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 218715000
Price (USD)/ Each
241.65
Online exclusive
284.00 
Save 42.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
241.65
Online exclusive
284.00 
Save 42.35 (15%)
Add to cart
Lanthanum(III) nitrate hexahydrate, 98+%, Thermo Scientific Chemicals
Catalog number218715000
Price (USD)/ Each
241.65
Online exclusive
284.00 
Save 42.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namelanthanum(3+) hexahydrate trinitrate
Molecular FormulaH12LaN3O15
InChI KeyGJKFIJKSBFYMQK-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol)433.01
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Appearance (Color)White
Titration Complexometric>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only