Sodium sulfate, 99+%, for analysis, powder, anhydrous, Thermo Scientific Chemicals
Sodium sulfate, 99+%, for analysis, powder, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfate, 99+%, for analysis, powder, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 218755000
Price (USD)/ Each
75.65
Online exclusive
83.70 
Save 8.05 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
75.65
Online exclusive
83.70 
Save 8.05 (10%)
Add to cart
Sodium sulfate, 99+%, for analysis, powder, anhydrous, Thermo Scientific Chemicals
Catalog number218755000
Price (USD)/ Each
75.65
Online exclusive
83.70 
Save 8.05 (10%)
-
Add to cart
Chemical Identifiers
CAS7757-82-6
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration after Ion exchange>=99.0 %
pH5 to 8 (5 % at 20°C)
Loss on ignition=<0.5 % (800°C)
Iron (Fe)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only