4-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals
4-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 218760050
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.80 
Save 8.15 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.80 
Save 8.15 (15%)
Add to cart
4-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals
Catalog number218760050
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.80 
Save 8.15 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-isocyanato-4-methoxybenzene
Molecular FormulaC8H7NO2
InChI KeyFMDGXCSMDZMDHZ-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)N=C=O
Molecular Weight (g/mol)149.15
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5450 to 1.5470 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only