Trifluoroacetic acid, ammonium salt, 98%, Thermo Scientific Chemicals
Trifluoroacetic acid, ammonium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetic acid, ammonium salt, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 218860250
Price (USD)/ Each
68.30
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
68.30
Add to cart
Trifluoroacetic acid, ammonium salt, 98%, Thermo Scientific Chemicals
Catalog number218860250
Price (USD)/ Each
68.30
-
Add to cart
Chemical Identifiers
IUPAC Nameammonium trifluoroacetic acid
Molecular FormulaC2H5F3NO2
InChI KeyYCNIBOIOWCTRCL-UHFFFAOYSA-O
SMILES[NH4+].OC(=O)C(F)(F)F
Molecular Weight (g/mol)132.06
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point122°C to 126°C
Titration with HClO4>=97.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only