Trifluoroacetic acid, ammonium salt, 98%, Thermo Scientific Chemicals
Trifluoroacetic acid, ammonium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetic acid, ammonium salt, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 218861000
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
Add to cart
Trifluoroacetic acid, ammonium salt, 98%, Thermo Scientific Chemicals
Catalog number218861000
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3336-58-1
IUPAC Nameammonium trifluoroacetic acid
Molecular FormulaC2H5F3NO2
InChI KeyYCNIBOIOWCTRCL-UHFFFAOYSA-O
SMILES[NH4+].OC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point122°C to 126°C
Titration with HClO4>=97.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only