(S)-(+)-1-Amino-2-propanol, 98+%, Thermo Scientific Chemicals
(S)-(+)-1-Amino-2-propanol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(+)-1-Amino-2-propanol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 218980250
Price (USD)/ Each
1,038.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
1,038.00
Add to cart
(S)-(+)-1-Amino-2-propanol, 98+%, Thermo Scientific Chemicals
Catalog number218980250
Price (USD)/ Each
1,038.00
-
Add to cart
Chemical Identifiers
CAS2799-17-9
IUPAC Name1-aminopropan-2-ol
Molecular FormulaC3H9NO
InChI KeyHXKKHQJGJAFBHI-UHFFFAOYNA-N
SMILESCC(O)CN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
Specific optical rotation+16° to +20° (20°C, 589 nm) (c=1.8, H2O)
Appearance (Color)Clear colorless to light yellow
GC>=98.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only