Thermo Scientific Chemicals

Methyl isocyanoacetate, 95%, Tech., Thermo Scientific Chemicals

Catalog number: 219000250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl isocyanoacetate, 95%, Tech., Thermo Scientific Chemicals

Catalog number: 219000250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 219000250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39687-95-1
IUPAC Name
methyl 2-(methylideneamino)acetate
Molecular Formula
C4H7NO2
InChI Key
ZRGVTTCCASYOFN-UHFFFAOYSA-N
SMILES
COC(=O)CN=C
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=94 %
Refractive index
1.4161 to 1.4191 (20°C, 589 nm)
Appearance (Color)
Clear yellow to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text