2-(Trimethylsilyl)ethoxymethyl chloride, 90%, tech., stabilized, Thermo Scientific Chemicals
2-(Trimethylsilyl)ethoxymethyl chloride, 90%, tech., stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Trimethylsilyl)ethoxymethyl chloride, 90%, tech., stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number 219020250
Price (USD)/ Each
524.65
Online exclusive
617.00 
Save 92.35 (15%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
524.65
Online exclusive
617.00 
Save 92.35 (15%)
Add to cart
2-(Trimethylsilyl)ethoxymethyl chloride, 90%, tech., stabilized, Thermo Scientific Chemicals
Catalog number219020250
Price (USD)/ Each
524.65
Online exclusive
617.00 
Save 92.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name[2-(chloromethoxy)ethyl]trimethylsilane
Molecular FormulaC6H15ClOSi
InChI KeyBPXKZEMBEZGUAH-UHFFFAOYSA-N
SMILESC[Si](C)(C)CCOCCl
Molecular Weight (g/mol)166.72
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=88 %
Stabilizer=<200 ppm Diisopropylethylamine
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only