Thermo Scientific Chemicals

2-(Trimethylsilyl)ethoxymethyl chloride, 90%, tech., stabilized, Thermo Scientific Chemicals

Catalog number: 219021000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-(Trimethylsilyl)ethoxymethyl chloride, 90%, tech., stabilized, Thermo Scientific Chemicals

Catalog number: 219021000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 219021000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
76513-69-4
IUPAC Name
[2-(chloromethoxy)ethyl]trimethylsilane
Molecular Formula
C6H15ClOSi
InChI Key
BPXKZEMBEZGUAH-UHFFFAOYSA-N
SMILES
C[Si](C)(C)CCOCCl
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
GC
>=88 %
Stabilizer
=<200 ppm Diisopropylethylamine
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text