Drierite™, without indicator, 6 mesh, Thermo Scientific Chemicals
Drierite™, without indicator, 6 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Drierite™, without indicator, 6 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number 219045000
Price (USD)/ Each
42.65
Online exclusive
49.90 
Save 7.25 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
42.65
Online exclusive
49.90 
Save 7.25 (15%)
Add to cart
Drierite™, without indicator, 6 mesh, Thermo Scientific Chemicals
Catalog number219045000
Price (USD)/ Each
42.65
Online exclusive
49.90 
Save 7.25 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namecalcium sulfate
Molecular FormulaCaO4S
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol)136.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Irregular granules
Composition>=99.5 % (CaSO4) Typical
Appearance (Color)White to greyish-white
Water=<0.3 %
Particle size6 mesh
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
RUO – Research Use Only