Drierite™, without indicator, 20-40 mesh, Thermo Scientific Chemicals
Drierite™, without indicator, 20-40 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Drierite™, without indicator, 20-40 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
454 g
2.3 kg
Packaging:
View all
Glass Bottle
Catalog number 219074540
Price (USD)/ Each
-
Quantity:
454 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namecalcium sulfate
Molecular FormulaCaO4S
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol)136.13
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.3 %
Particle size20 to 40 mesh
Appearance (Form)Irregular granules
Composition>=99.5 % (CaSO4) Typical
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
RUO – Research Use Only