Drierite™, with indicator, 8 mesh
Drierite™, with indicator, 8 mesh
Drierite™, with indicator, 8 mesh
Drierite™, with indicator, 8 mesh
Thermo Scientific Chemicals

Drierite™, with indicator, 8 mesh

Drierite, CaO4S, CAS Number-7778-18-9, 7646-79-9, 5kg, 39.09%, 0.68%, CHEBI:31346, 3.25%, Blue, 01,107, 136.13, GHS Signal Word: Danger | CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
2 kg
Packaging:
View all
Glass Bottle
Catalog number 219090020
Price (USD)
219.65
Special offer
Online exclusive
Ends: 26-Sep-2025
271.00
Save 51.35 (19%)
Each
-
Add to cart
Quantity:
2 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
219.65
Special offer
Online exclusive
Ends: 26-Sep-2025
271.00
Save 51.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS7778-18-9
IUPAC Namecalcium sulfate
Molecular FormulaCaO4S
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue
Appearance (Form)Irregular granules
Water=<0.3 %
Composition>=98 % (CaSO4) Typical
Composition=<2 % (CoCl2) Typical
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection