Drierite, with indicator, 8 mesh, Thermo Scientific Chemicals
Drierite, with indicator, 8 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Drierite, with indicator, 8 mesh, Thermo Scientific Chemicals

Drierite, CaO4S, CAS Number-7778-18-9, 7646-79-9, 5kg, 39.09%, 0.68%, CHEBI:31346, 3.25%, Blue, 01,107, 136.13, GHS Signal Word: Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
5 kg
Packaging:
View all
Glass Bottle
Catalog number 219095000
Price (USD)/ Each
84.65
Online exclusive
94.40 
Save 9.75 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
84.65
Online exclusive
94.40 
Save 9.75 (10%)
Add to cart
Drierite, with indicator, 8 mesh, Thermo Scientific Chemicals
Catalog number219095000
Price (USD)/ Each
84.65
Online exclusive
94.40 
Save 9.75 (10%)
-
Add to cart
Chemical Identifiers
CAS7778-18-9
IUPAC Namecalcium sulfate
Molecular FormulaCaO4S
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue
Water=<0.3 %
Composition>=98 % (CaSO4) Typical
Particle size8 mesh
Appearance (Form)Irregular granules
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
RUO – Research Use Only