Thermo Scientific Chemicals

Cesium, 99.95+%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 219190010
1 g, Each
Thermo Scientific Chemicals

Cesium, 99.95+%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 219190010
1 g, Each
Quantity
Catalog number: 219190010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-46-2
IUPAC Name
caesium
Molecular Formula
Cs
InChI Key
TVFDJXOCXUVLDH-UHFFFAOYSA-N
SMILES
[Cs]
Appearance (Form)
Metal
Appearance (Color)
Golden
Cesium (Cs)
>=99.95 % (metals basis)
Total trace metal impurities
=<500 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text