Chromium(III) nitrate nonahydrate, 99%
Chromium(III) nitrate nonahydrate, 99%
Chromium(III) nitrate nonahydrate, 99%
Chromium(III) nitrate nonahydrate, 99%
Thermo Scientific Chemicals

Chromium(III) nitrate nonahydrate, 99%

CAS: 7789-02-8 | CrN3O9·9H2O | 400.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 219201000
Price (USD)
47.65
Special offer
Online exclusive
Ends: 26-Sep-2025
58.10
Save 10.45 (18%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 26-Sep-2025
58.10
Save 10.45 (18%)
Each
Add to cart
Chemical Identifiers
CAS7789-02-8
IUPAC Namechromium(3+) trinitrate
Molecular FormulaCrN3O9
InChI KeyPHFQLYPOURZARY-UHFFFAOYSA-N
SMILES[Cr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Calcium (Ca)=<50 ppm
Copper (Cu)=<10 ppm
Magnesium (Mg)=<50 ppm
Sodium (Na)=<50 ppm
Appearance (Form)Crystalline chunks
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link