Thermo Scientific Chemicals

Potassium hydrogen fluoride, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 219230250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium hydrogen fluoride, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 219230250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 219230250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-29-9
IUPAC Name
potassium hydrogen difluoride
Molecular Formula
F2HK
InChI Key
VBKNTGMWIPUCRF-UHFFFAOYSA-M
SMILES
[H+].[F-].[F-].[K+]
Appearance (Color)
White to light grey
Appearance (Form)
Crystalline powder or crystals
Titration with NaOH
99.0 to 101.0 %
Water
=<0.3 % (KF)
Heavy metals (as Pb)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text