Sodium sulfate, 99+%, for analysis, crystalline, anhydrous, Thermo Scientific Chemicals
Sodium sulfate, 99+%, for analysis, crystalline, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfate, 99+%, for analysis, crystalline, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 219260100
Price (USD)/ Each
-
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
Molecular Weight (g/mol)142.04
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Magnesium (Mg)=<50 ppm
Loss on ignition=<0.5 %
Phosphate (PO4)=<20 ppm
Potassium (K)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only