Sodium sulfite, 98.5%, for analysis, anhydrous, Thermo Scientific Chemicals
Sodium sulfite, 98.5%, for analysis, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfite, 98.5%, for analysis, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
View all
Glass Bottle
Catalog number 219270250
Price (USD)/ Each
25.65
Online exclusive
30.40 
Save 4.75 (16%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
25.65
Online exclusive
30.40 
Save 4.75 (16%)
Add to cart
Sodium sulfite, 98.5%, for analysis, anhydrous, Thermo Scientific Chemicals
Catalog number219270250
Price (USD)/ Each
25.65
Online exclusive
30.40 
Save 4.75 (16%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium heptahydrate sulfite
Molecular FormulaH14Na2O10S
InChI KeyWMJRPJZQQSSDBU-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O
Molecular Weight (g/mol)252.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm
Appearance (Form)Crystalline powder or crystals
Titration Iodimetric>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only