Thermo Scientific Chemicals

2-Ethylimidazole, 99%, Thermo Scientific Chemicals

Catalog number: 219285000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Ethylimidazole, 99%, Thermo Scientific Chemicals

Catalog number: 219285000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 219285000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1072-62-4
IUPAC Name
2-ethyl-1H-imidazole
Molecular Formula
C5H8N2
InChI Key
PQAMFDRRWURCFQ-UHFFFAOYSA-N
SMILES
CCC1=NC=CN1
Appearance (Color)
White to light yellow
Appearance (Form)
Flakes
GC
>=98.5 %
Melting point
83°C to 87°C
GC
=<1.0 % Imidazole

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text