2-Formylbenzenesulfonic acid sodium salt, 90%, Tech., Thermo Scientific Chemicals
2-Formylbenzenesulfonic acid sodium salt, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Formylbenzenesulfonic acid sodium salt, 90%, Tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number 219320025
Price (USD)/ Each
574.00
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
574.00
Add to cart
2-Formylbenzenesulfonic acid sodium salt, 90%, Tech., Thermo Scientific Chemicals
Catalog number219320025
Price (USD)/ Each
574.00
-
Add to cart
Chemical Identifiers
CAS1008-72-6
IUPAC Namesodium 2-formylbenzene-1-sulfonate
Molecular FormulaC7H5NaO4S
InChI KeyADPUQRRLAAPXGT-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with NaOH>=88 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Sodium chloride (NaCl)=<3 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only