Thermo Scientific Chemicals

2-Formylbenzenesulfonic acid sodium salt, 90%, Tech., Thermo Scientific Chemicals

Catalog number: 219325000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-Formylbenzenesulfonic acid sodium salt, 90%, Tech., Thermo Scientific Chemicals

Catalog number: 219325000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 219325000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1008-72-6
IUPAC Name
sodium 2-formylbenzene-1-sulfonate
Molecular Formula
C7H5NaO4S
InChI Key
ADPUQRRLAAPXGT-UHFFFAOYSA-M
SMILES
[Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Sodium chloride (NaCl)
=<3 %
Appearance (Form)
Powder
Titration with NaOH
>=88 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text